Merck
  • The metabolic sensor PASK is a histone 3 kinase that also regulates H3K4 methylation by associating with H3K4 MLL2 methyltransferase complex.
The metabolic sensor PASK is a histone 3 kinase that also regulates H3K4 methylation by associating with H3K4 MLL2 methyltransferase complex.
Nucleic acids research (2019-09-19)
Jimsheena V Karakkat, Suneesh Kaimala, Sreejisha P Sreedharan, Princy Jayaprakash, Ernest A Adeghate, Suraiya A Ansari, Ernesto Guccione, Eric P K Mensah-Brown, Bright Starling Emerald
ABSTRACT
The metabolic sensor Per-Arnt-Sim (Pas) domain-containing serine/threonine kinase (PASK) is expressed predominantly in the cytoplasm of different cell types, although a small percentage is also expressed in the nucleus. Herein, we show that the nuclear PASK associates with the mammalian H3K4 MLL2 methyltransferase complex and enhances H3K4 di- and tri-methylation. We also show that PASK is a histone kinase that phosphorylates H3 at T3, T6, S10 and T11. Taken together, these results suggest that PASK regulates two different H3 tail modifications involving H3K4 methylation and H3 phosphorylation. Using muscle satellite cell differentiation and functional analysis after loss or gain of Pask expression using the CRISPR/Cas9 system, we provide evidence that some of the regulatory functions of PASK during development and differentiation may occur through the regulation of these histone modifications.
Product Description
Millipore
Anti-V5 Agarose Affinity Gel antibody produced in mouse, purified immunoglobulin, clone V5-10
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-monomethyl Histone H3 (Lys4) Antibody, clone RM140, clone RM140, from rabbit
Sigma-Aldrich
Hydroxyurea, Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase.