Merck
  • The human Shu complex functions with PDS5B and SPIDR to promote homologous recombination.
The human Shu complex functions with PDS5B and SPIDR to promote homologous recombination.
Nucleic acids research (2019-10-31)
Julieta Martino, Gregory J Brunette, Jonathan Barroso-González, Tatiana N Moiseeva, Chelsea M Smith, Christopher J Bakkenist, Roderick J O'Sullivan, Kara A Bernstein
ABSTRACT
RAD51 plays a central role in homologous recombination during double-strand break repair and in replication fork dynamics. Misregulation of RAD51 is associated with genetic instability and cancer. RAD51 is regulated by many accessory proteins including the highly conserved Shu complex. Here, we report the function of the human Shu complex during replication to regulate RAD51 recruitment to DNA repair foci and, secondly, during replication fork restart following replication fork stalling. Deletion of the Shu complex members, SWS1 and SWSAP1, using CRISPR/Cas9, renders cells specifically sensitive to the replication fork stalling and collapse caused by methyl methanesulfonate and mitomycin C exposure, a delayed and reduced RAD51 response, and fewer sister chromatid exchanges. Our additional analysis identified SPIDR and PDS5B as novel Shu complex interacting partners and genetically function in the same pathway upon DNA damage. Collectively, our study uncovers a protein complex, which consists of SWS1, SWSAP1, SPIDR and PDS5B, involved in DNA repair and provides insight into Shu complex function and composition.
Sigma-Aldrich
Demecolcine solution, 10 μg/mL in HBSS, ACF Qualified, BioXtra
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Anti-SPIDR antibody produced in rabbit, affinity isolated antibody
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Supelco
Methyl methanesulfonate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Methyl methanesulfonate, 99%
Millipore
Benzonase® Nuclease, Purity > 90%