Merck
  • Weak Fragment Crystallizable (Fc) Domain Interactions Drive the Dynamic Assembly of IgG Oligomers upon Antigen Recognition.
Weak Fragment Crystallizable (Fc) Domain Interactions Drive the Dynamic Assembly of IgG Oligomers upon Antigen Recognition.
ACS nano (2019-12-31)
Jürgen Strasser, Rob N de Jong, Frank J Beurskens, Janine Schuurman, Paul W H I Parren, Peter Hinterdorfer, Johannes Preiner
ABSTRACT
Activation of membrane receptors through clustering is a common mechanism found in various biological systems. Spatial proximity of receptors may be transduced across the membrane to initiate signaling pathways or alternatively be recognized by peripheral proteins or immune cells to trigger external effector functions. Here we show how specific immunoglobulin G (IgG) binding induces clustering of monomeric target molecules in lipid membranes through Fc-Fc interactions. We visualize and characterize the dynamic IgG oligomerization process and the molecular interactions involved using high-speed atomic force microscopy, single-molecule force spectroscopy, and quartz crystal microbalance experiments. We found that the Fc-Fc interaction strength is precisely tuned to be weak enough to prevent IgG oligomerization in solution at physiological titers, but enabling IgG oligomerization when Fabs additionally bind to their cognate surface epitopes, a mechanism that ultimately targets IgG-mediated effector functions such as classical complement activation to antigenic membranes.
Product Description
Avanti
16:0 DNP Cap PE, 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[6-[(2,4-dinitrophenyl)amino]hexanoyl] (ammonium salt), powder
Avanti
16:0 DNP Cap PE, 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[6-[(2,4-dinitrophenyl)amino]hexanoyl] (ammonium salt), chloroform
Avanti
16:0 PE, Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand