Merck
  • Recombinant hepatitis C virus genotype 5a infectious cell culture systems expressing minimal JFH1 NS5B sequences permit polymerase inhibitor studies.
Recombinant hepatitis C virus genotype 5a infectious cell culture systems expressing minimal JFH1 NS5B sequences permit polymerase inhibitor studies.
Virology (2018-07-23)
Daryl Humes, Santseharay Ramirez, Tanja B Jensen, Yi-Ping Li, Judith M Gottwein, Jens Bukh
ABSTRACT
The six major epidemiologically important hepatitis C virus (HCV) genotypes differ in global distribution and antiviral responses. Full-length infectious cell-culture adapted clones, the gold standard for HCV studies in vitro, are missing for genotypes 4 and 5. To address this challenge for genotype 5, we constructed a consensus full-length clone of strain SA13 (SA13fl), which was found non-viable in Huh7.5 cells. Step-wise adaptation of SA13fl-based recombinants, beginning with a virus encoding the NS5B-thumb domain and 3´UTR of JFH1 (SA13/JF372-X), resulted in a high-titer SA13 virus with only 41 JFH1-encoded NS5B-thumb residues (SA13/JF470-510cc); this required sixteen cell-culture adaptive substitutions within the SA13fl polyprotein and two 3´UTR-changes. SA13/JF372-X and SA13/JF470-510cc were equally sensitive to nucleoside polymerase inhibitors, including sofosbuvir, but showed differential sensitivity to inhibitors targeting the NS5B palm or thumb. SA13/JF470-510cc represents a model to elucidate the influence of HCV RNA elements on viral replication and map determinants of sensitivity to polymerase inhibitors.
Sigma-Aldrich
Lomibuvir, ≥98% (HPLC)