Merck
  • Intramolecular Ring-Expansion Reaction (RER) and Intermolecular Coordination of In Situ Generated Cyclic (Amino)(aryl)carbenes (cAArCs).
Intramolecular Ring-Expansion Reaction (RER) and Intermolecular Coordination of In Situ Generated Cyclic (Amino)(aryl)carbenes (cAArCs).
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-07-06)
Jan Lorkowski, Mirjam Krahfuß, Maciej Kubicki, Udo Radius, Cezary Pietraszuk
ABSTRACT
Cyclic (amino)(aryl)carbenes (cAArCs) based on the isoindoline core were successfully generated in situ by α-elimination of 3-alkoxyisoindolines at high temperatures or by deprotonation of isoindol-2-ium chlorides with sodium or copper(I) acetates at low temperatures. 3-Alkoxy-isoindolines 2 a,b-OR (R=Me, Et, iPr) have been prepared in high yields by the addition of a solution of 2-aryl-1,1-diphenylisoindol-2-ium triflate (1 a,b-OTf; a: aryl=Dipp=2,6-diisopropylphenyl; b: Mesityl-, Mes=2,4,6-trimethylphenyl) to the corresponding alcohol (ROH) with NEt3 at room temperature. Furthermore, the reaction of 2 a,b-OMe in diethyl ether with a tenfold excess of hydrochloric acid led to the isolation of the isoindol-2-ium chlorides 1 a,b-Cl in high yields. The thermally generated cAArC reacts with sulfur to form the thioamide 3 a. Without any additional trapping reagent, in situ generation of 1,1-diphenylisoidolin-3-ylidenes does not lead to the isolation of these compounds, but to the reaction products of the insertion of the carbene carbon atom into an ortho C-H bond of a phenyl substituent, followed by ring-expansion reaction; namely, anthracene derivatives 9-N(H)aryl-10-Ph-C14 H8 4 a,b (a: Dipp; b: Mes). These compounds are conveniently synthesized by deprotonation of the isoindol-2-ium chlorides with sodium acetate in high yields. Deprotonation of 1 a-Cl with copper(I) acetate at low temperatures afforded a mixture of 4 a and the corresponding cAArC copper(I) chloride 5 a, and allowed the isolation and structural characterization of the first example of a cAArC copper complex of general formula [(cAArC)CuCl].
Sigma-Aldrich
Copper(I) acetate, 97%