Merck
  • Sirtuin 7 promotes 45S pre-rRNA cleavage at site 2 and determines the processing pathway.
Sirtuin 7 promotes 45S pre-rRNA cleavage at site 2 and determines the processing pathway.
Journal of cell science (2019-07-25)
Valentina Sirri, Alice Grob, Jérémy Berthelet, Nathalie Jourdan, Pascal Roussel
ABSTRACT
In humans, ribosome biogenesis mainly occurs in nucleoli following two alternative pre-rRNA processing pathways differing in the order in which cleavages take place but not by the sites of cleavage. To uncover the role of the nucleolar NAD+-dependent deacetylase sirtuin 7 in the synthesis of ribosomal subunits, pre-rRNA processing was analyzed after sirtinol-mediated inhibition of sirtuin 7 activity or depletion of sirtuin 7 protein. We thus reveal that sirtuin 7 activity is a critical regulator of processing of 45S, 32S and 30S pre-rRNAs. Sirtuin 7 protein is primarily essential to 45S pre-rRNA cleavage at site 2, which is the first step of processing pathway 2. Furthermore, we demonstrate that sirtuin 7 physically interacts with Nop56 and the GAR domain of fibrillarin, and propose that this could interfere with fibrillarin-dependent cleavage. Sirtuin 7 depletion results in the accumulation of 5' extended forms of 32S pre-rRNA, and also influences the localization of fibrillarin. Thus, we establish a close relationship between sirtuin 7 and fibrillarin, which might determine the processing pathway used for ribosome biogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human FBL
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Millipore
Glutathione−Agarose, lyophilized powder
Sigma-Aldrich
Anti-Sirt7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SIRT7
Sigma-Aldrich
MISSION® esiRNA, targeting human NCL
Sigma-Aldrich
SIRT7, active, His tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution