Merck
  • Unfolded protein response regulates P53 expression in the pulmonary endothelium.
Unfolded protein response regulates P53 expression in the pulmonary endothelium.
Journal of biochemical and molecular toxicology (2019-07-25)
Mohammad S Akhter, Mohammad A Uddin, Nektarios Barabutis
ABSTRACT
Lung endothelial barrier dysfunction leads to severe pathologies, including the lethal Acute Respiratory Distress Syndrome. P53 has been associated with anti-inflammatory activities. The current study employs a variety of unfolded protein response (UPR) activators and inhibitors to investigate the regulation of P53 by UPR in lung cells. The bovine cells that were exposed to the UPR inductors brefeldin A, dithiothreitol, and thapsigargin; demonstrated elevated expression levels of P53 compared to the vehicle-treated cells. On the contrary, the UPR inhibitors N-acetyl cysteine, kifunensine, and ATP-competitive IRE1α kinase-inhibiting RNase attenuator; produced the opposite effects. The outcomes of the present study reveal a positive regulation between UPR and P53. Since it has been shown that a mild induction of the unfolded protein response opposes inflammation, we suggest that P53 is involved in those protective activities in the lung.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
IRE1 Inhibitor IV, KIRA6
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)