Merck
  • In Vivo CRISPR Screen Identifies TgWIP as a Toxoplasma Modulator of Dendritic Cell Migration.
In Vivo CRISPR Screen Identifies TgWIP as a Toxoplasma Modulator of Dendritic Cell Migration.
Cell host & microbe (2019-10-11)
Lamba Omar Sangaré, Einar B Ólafsson, Yifan Wang, Ninghan Yang, Lindsay Julien, Ana Camejo, Patricia Pesavento, Saima M Sidik, Sebastian Lourido, Antonio Barragan, Jeroen P J Saeij
ABSTRACT
Toxoplasma can reach distant organs, especially the brain, leading to a lifelong chronic phase. However, genes involved in related in vivo processes are currently unknown. Here, we use focused CRISPR libraries to identify Toxoplasma genes that affect in vivo fitness. We focus on TgWIP, whose deletion affects Toxoplasma dissemination to distant organs. We show that TgWIP is secreted into the host cell upon invasion and interacts with the host WAVE regulatory complex and SHP2 phosphatase, both of which regulate actin dynamics. TgWIP affects the morphology of dendritic cells and mediates the dissolution of podosomes, which dendritic cells use to adhere to extracellular matrix. TgWIP enhances the motility and transmigration of parasitized dendritic cells, likely explaining its effect on in vivo fitness. Our results provide a framework for systemic identification of Toxoplasma genes with in vivo effects at the site of infection or on dissemination to distant organs, including the brain.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Pricing and availability is not currently available.
Supelco
Mycophenolic acid, analytical standard
Pricing and availability is not currently available.
Corning® cell strainer, pore size 70 μm, white, sterile, pkg of (individually wrapped), pack of 50 ea, Corning 431751
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.