Merck
  • JLP-centrosome is essential for the microtubule-mediated nucleocytoplasmic transport induced by extracellular stimuli.
JLP-centrosome is essential for the microtubule-mediated nucleocytoplasmic transport induced by extracellular stimuli.
Science advances (2019-12-06)
Clement M Lee, Ken Aizawa, Joshua Jiang, Sam K P Kung, Rinku Jain
ABSTRACT
JLP belongs to the JIP family whose members serve as scaffolding proteins that link motor proteins and their cargo for intracellular transport. Although JLP is mainly cytoplasmic, it accumulates as a focus in the perinuclear region when stimulated by extracellular stimuli. Focus formation, which changes the nucleus shape and concentrates the nuclear pores, depends on p38MAPK activation and the dynein retrograde motor protein complex. Extracellular stimuli trigger the tethering of PLK1 to the centrosome by JLP, leading to centrosome maturation and microtubule array formation. The centrosome localization domain of JLP is important for the binding of the centrosome and the formation of the JLP focus and the microtubule array. Furthermore, the formation of the JLP focus and the microtubule array is interdependent and important for the transport of NF-κB p65 to the nucleus and its unloading therein. In conclusion, JLP exhibits multiple functions in the nuclear translocation of NF-κB p65.
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Rose Bengal lactone, 95%
Sigma-Aldrich
PD 169316, ≥98% (HPLC), solid
Millipore
S-protein Agarose
Millipore
Glutathione−Agarose, lyophilized powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
EHNA hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).