Merck
  • Lipid droplet size directs lipolysis and lipophagy catabolism in hepatocytes.
Lipid droplet size directs lipolysis and lipophagy catabolism in hepatocytes.
The Journal of cell biology (2019-08-09)
Micah B Schott, Shaun G Weller, Ryan J Schulze, Eugene W Krueger, Kristina Drizyte-Miller, Carol A Casey, Mark A McNiven
ABSTRACT
Lipid droplet (LD) catabolism in hepatocytes is mediated by a combination of lipolysis and a selective autophagic mechanism called lipophagy, but the relative contributions of these seemingly distinct pathways remain unclear. We find that inhibition of lipolysis, lipophagy, or both resulted in similar overall LD content but dramatic differences in LD morphology. Inhibition of the lipolysis enzyme adipose triglyceride lipase (ATGL) resulted in large cytoplasmic LDs, whereas lysosomal inhibition caused the accumulation of numerous small LDs within the cytoplasm and degradative acidic vesicles. Combined inhibition of ATGL and LAL resulted in large LDs, suggesting that lipolysis targets these LDs upstream of lipophagy. Consistent with this, ATGL was enriched in larger-sized LDs, whereas lipophagic vesicles were restricted to small LDs as revealed by immunofluorescence, electron microscopy, and Western blot of size-separated LDs. These findings provide new evidence indicating a synergistic relationship whereby lipolysis targets larger-sized LDs to produce both size-reduced and nascently synthesized small LDs that are amenable for lipophagic internalization.
Sigma-Aldrich
Lipoprotein Deficient Serum from fetal calf, frozen liquid
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%