Merck
  • Precise Positioning of Organic Semiconductor Single Crystals with Two-Component Aligned Structure through 3D Wettability-Induced Sequential Assembly.
Precise Positioning of Organic Semiconductor Single Crystals with Two-Component Aligned Structure through 3D Wettability-Induced Sequential Assembly.
ACS applied materials & interfaces (2019-08-31)
Wei Deng, Bei Lu, Jian Mao, Zhengjun Lu, Xiujuan Zhang, Jiansheng Jie
ABSTRACT
Highly ordered organic semiconductor single-crystal (OSSC) arrays are ideal building blocks for functional organic devices. However, most of the current methods are only applicable to fabricate OSSC arrays of a single component, which significantly hinders the application of OSSC arrays in integrated organic circuits. Here, we present a universal approach, termed three-dimensional (3D) wettability-induced sequential assembly that can programmatically and progressively manipulate the crystallization locations of different organic semiconductors at the same spatial position using a 3D microchannel template, for the fabrication of the two-component OSSC arrays. As an example, we successfully prepared two-component, bilayer structured OSSC arrays consisting of n-type N,N'-bis(2-phenylethyl)-perylene-3,4:9,10-tetracarboxylic diimide and p-type 6,13-bis(triisopropylsilylethynyl)pentacene microbelts. The bicomponent OSSCs show ambipolar carrier transport properties with hole and electron mobilities of 0.342 and 0.526 cm2 V-1 s-1, respectively. Construction of complementary inverters is further demonstrated based on the two-component OSSCs. The capability of integration of multicomponent OSSC arrays opens up unique opportunities for future high-performance organic complementary circuits.
Sigma-Aldrich
N,N′-Dioctyl-3,4,9,10-perylenedicarboximide, 98%