Merck
  • Growth/differentiation 5 promotes the differentiation of retinal stem cells into neurons via Atoh8.
Growth/differentiation 5 promotes the differentiation of retinal stem cells into neurons via Atoh8.
Journal of cellular physiology (2019-05-09)
Yanxiu Li, Haibo Li, LuSi Zhang, Siqi Xiong, ShiJin Wen, Xiaobo Xia, Xia Zhou
ABSTRACT
Retinal diseases are characterized by the degeneration of retinal neural cells, and are the main cause of blindness. Although the development of stem cell including retinal stem cell therapies raises hope for retinal neuron replacement, currently, there is still no efficient method to regenerate retinal neurons. To realize the potential roles of the production of retinal neurons, neurotrophic factor direct the differentiation of retinal stem cells should be extensively identified. In this article, we characterized growth/differentiation 5 (GDF5), which caused the activation of Smad signaling, can induce neurogenesis and neurite outgrowth in retinal stem cell differentiation. Moreover, a bHLH transcription factor, Atoh8 modulates the effects stimulated by GDF5. These data suggested that GDF5 regulates neuron differentiation through mediating Atoh8 and help us to understand the pathophysiological function of GDF5 in retinal regeneration.
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Nestin (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
Sigma-Aldrich
LDN193189 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
SB-505124 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Anti-ATOH8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-SOX2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL4716, purified immunoglobulin