Merck
  • The Cardiac Microenvironment Instructs Divergent Monocyte Fates and Functions in Myocarditis.
The Cardiac Microenvironment Instructs Divergent Monocyte Fates and Functions in Myocarditis.
Cell reports (2019-07-04)
Xuezhou Hou, Guobao Chen, William Bracamonte-Baran, Hee Sun Choi, Nicola L Diny, Jungeun Sung, David Hughes, Taejoon Won, Megan Kay Wood, Monica V Talor, David Joel Hackam, Karin Klingel, Giovanni Davogustto, Heinrich Taegtmeyer, Isabelle Coppens, Jobert G Barin, Daniela Čiháková
ABSTRACT
Two types of monocytes, Ly6Chi and Ly6Clo, infiltrate the heart in murine experimental autoimmune myocarditis (EAM). We discovered a role for cardiac fibroblasts in facilitating monocyte-to-macrophage differentiation of both Ly6Chi and Ly6Clo cells, allowing these macrophages to perform divergent functions in myocarditis progression. During the acute phase of EAM, IL-17A is highly abundant. It signals through cardiac fibroblasts to attenuate efferocytosis of Ly6Chi monocyte-derived macrophages (MDMs) and simultaneously prevents Ly6Clo monocyte-to-macrophage differentiation. We demonstrated an inverse clinical correlation between heart IL-17A levels and efferocytic receptor expressions in humans with heart failure (HF). In the absence of IL-17A signaling, Ly6Chi MDMs act as robust phagocytes and are less pro-inflammatory, whereas Ly6Clo monocytes resume their differentiation into MHCII+ macrophages. We propose that MHCII+Ly6Clo MDMs are associated with the reduction of cardiac fibrosis and prevention of the myocarditis sequalae.
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%