Merck
  • Junctional Adhesion Molecule 2 Represents a Subset of Hematopoietic Stem Cells with Enhanced Potential for T Lymphopoiesis.
Junctional Adhesion Molecule 2 Represents a Subset of Hematopoietic Stem Cells with Enhanced Potential for T Lymphopoiesis.
Cell reports (2019-06-06)
Visnja Radulovic, Mark van der Garde, Shuhei Koide, Valgardur Sigurdsson, Stefan Lang, Shin Kaneko, Kenichi Miharada
ABSTRACT
The distinct lineage potential is a key feature of hematopoietic stem cell (HSC) heterogeneity, but a subset of HSCs specialized for a single lymphoid compartment has not been identified. Here we report that HSCs expressing junctional adhesion molecule 2 (Jam2) at a higher level (Jam2high HSCs) have a greater T cell reconstitution capacity. Jam2high HSCs are metabolically dormant but preferentially differentiate toward lymphocytes, especially T cell lineages. Jam2high HSCs uniquely express T cell-related genes, and the interaction with Jam1 facilitates the Notch/Delta signaling pathway. Frequency of Jam2high HSCs changes upon T cell depletion in vivo, potentially suggesting that Jam2 expression may reflect scarcity of T cells and requirement of T cell replenishment. Our findings highlight Jam2 as a potential marker for a subfraction of HSCs with an extensive lymphopoietic capacity, mainly in T lymphopoiesis.