Merck
  • Temperature-Mediated Variations in Behavior and Mortality Caused by Non-Repellent Insecticides in Subterranean Termites (Blattodea: Rhinotermitidae).
Temperature-Mediated Variations in Behavior and Mortality Caused by Non-Repellent Insecticides in Subterranean Termites (Blattodea: Rhinotermitidae).
Insects (2019-02-02)
Franklin Y Quarcoo, Xing Ping Hu, Arthur G Appel
ABSTRACT
Behavioral symptoms and mortality associated with intoxication with insecticides fipronil and indoxacarb were determined in field-collected eastern subterranean termites, Reticulitermes flavipes (Kollar), and Formosan subterranean termites, Coptotermes formosanus Shiraki. Behaviors and mortality were evaluated at three temperatures (16, 22, and 28 °C) and three concentrations of fipronil (0.5, 1, and 5 ppm) and indoxacarb (50, 75, and 100 ppm). LT50 (median lethal time to kill 50% of the termites) values declined with increasing concentrations and temperatures for both fipronil-exposed eastern and Formosan subterranean termites, whereas these values were not always the highest at 16 °C for indoxacarb-treated termites. The greatest change (reduction) in LT50 values occurred for both species between 16 and 22 °C at the lowest concentration of each insecticide. Intoxication and moribundity were the most frequently observed behaviors for fipronil-exposed termites, whereas intoxication, ataxia, and moribundity were observed for most concentration and temperature combinations for indoxacarb-exposed termites. The inherent toxicity of fipronil was higher than that of indoxacarb. The higher presence and duration of intoxication behaviors may positively affect the performance of indoxacarb against subterranean termite colonies.