Merck
  • EZH2-Mediated Primary Cilium Deconstruction Drives Metastatic Melanoma Formation.
EZH2-Mediated Primary Cilium Deconstruction Drives Metastatic Melanoma Formation.
Cancer cell (2018-07-17)
Daniel Zingg, Julien Debbache, Rodrigo Peña-Hernández, Ana T Antunes, Simon M Schaefer, Phil F Cheng, Dario Zimmerli, Jessica Haeusel, Raquel R Calçada, Eylul Tuncer, Yudong Zhang, Raphaël Bossart, Kwok-Kin Wong, Konrad Basler, Reinhard Dummer, Raffaella Santoro, Mitchell P Levesque, Lukas Sommer
ABSTRACT
Human melanomas frequently harbor amplifications of EZH2. However, the contribution of EZH2 to melanoma formation has remained elusive. Taking advantage of murine melanoma models, we show that EZH2 drives tumorigenesis from benign BrafV600E- or NrasQ61K-expressing melanocytes by silencing of genes relevant for the integrity of the primary cilium, a signaling organelle projecting from the surface of vertebrate cells. Consequently, gain of EZH2 promotes loss of primary cilia in benign melanocytic lesions. In contrast, blockade of EZH2 activity evokes ciliogenesis and cilia-dependent growth inhibition in malignant melanoma. Finally, we demonstrate that loss of cilia enhances pro-tumorigenic WNT/β-catenin signaling, and is itself sufficient to drive metastatic melanoma in benign cells. Thus, primary cilia deconstruction is a key process in EZH2-driven melanomagenesis.
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Donkey Anti-Chicken IgY Antibody, biotin conjugate, Species Adsorbed, Chemicon®, from donkey