Merck
UBTD1 is a mechano-regulator controlling cancer aggressiveness.
EMBO reports (2019-02-26)
Stéphanie Torrino, François-René Roustan, Lisa Kaminski, Thomas Bertero, Sabrina Pisano, Damien Ambrosetti, Maeva Dufies, Jay P Uhler, Emmanuel Lemichez, Amel Mettouchi, Maeva Gesson, Kathiane Laurent, Cedric Gaggioli, Jean-Francois Michiels, Christophe Lamaze, Frédéric Bost, Stéphan Clavel
ABSTRACT
Ubiquitin domain-containing protein 1 (UBTD1) is highly evolutionary conserved and has been described to interact with E2 enzymes of the ubiquitin-proteasome system. However, its biological role and the functional significance of this interaction remain largely unknown. Here, we demonstrate that depletion of UBTD1 drastically affects the mechanical properties of epithelial cancer cells via RhoA activation and strongly promotes their aggressiveness. On a stiff matrix, UBTD1 expression is regulated by cell-cell contacts, and the protein is associated with β-catenin at cell junctions. Yes-associated protein (YAP) is a major cell mechano-transducer, and we show that UBTD1 is associated with components of the YAP degradation complex. Interestingly, UBTD1 promotes the interaction of YAP with its E3 ubiquitin ligase β-TrCP Consequently, in cancer cells, UBTD1 depletion decreases YAP ubiquitylation and triggers robust ROCK2-dependent YAP activation and downstream signaling. Data from lung and prostate cancer patients further corroborate the in cellulo results, confirming that low levels of UBTD1 are associated with poor patient survival, suggesting that biological functions of UBTD1 could be beneficial in limiting cancer progression.
Sigma-Aldrich
Anti-UBTD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)