Merck
  • Limbic Neurons Shape Sex Recognition and Social Behavior in Sexually Naive Males.
Limbic Neurons Shape Sex Recognition and Social Behavior in Sexually Naive Males.
Cell (2019-02-05)
Daniel W Bayless, Taehong Yang, Matthew M Mason, Albert A T Susanto, Alexandra Lobdell, Nirao M Shah
ABSTRACT
Sexually naive animals have to distinguish between the sexes because they show species-typical interactions with males and females without meaningful prior experience. However, central neural pathways in naive mammals that recognize sex of other individuals remain poorly characterized. We examined the role of the principal component of the bed nucleus of stria terminalis (BNSTpr), a limbic center, in social interactions in mice. We find that activity of aromatase-expressing BNSTpr (AB) neurons appears to encode sex of other animals and subsequent displays of mating in sexually naive males. Silencing these neurons in males eliminates preference for female pheromones and abrogates mating success, whereas activating them even transiently promotes male-male mating. Surprisingly, female AB neurons do not appear to control sex recognition, mating, or maternal aggression. In summary, AB neurons represent sex of other animals and govern ensuing social behaviors in sexually naive males.