Merck
  • Stereoselective synthesis of substituted 1,3-oxazolidines via Pd-catalyzed carboamination reactions of O-vinyl-1,2-amino alcohols.
Stereoselective synthesis of substituted 1,3-oxazolidines via Pd-catalyzed carboamination reactions of O-vinyl-1,2-amino alcohols.
Organic letters (2011-08-05)
Amanda F Ward, John P Wolfe
ABSTRACT
The stereoselective synthesis of 2,4- and 2,5-disubstituted 1,3-oxazolidines is accomplished via Pd-catalyzed carboamination of O-vinyl-1,2-amino alcohol derivatives. The transformations generate cis-disubstituted products with good to excellent diastereoselectivity, and enantiomerically enriched substrates are converted without loss of optical purity. In addition to yielding synthetically useful products that are difficult to generate with existing methods, these transformations illustrate that electron-rich enol ethers are viable substrates for alkene carboamination processes.
Sigma-Aldrich
Vinylboronic anhydride pyridine complex, 95%