Merck
Study of thiol proteases of normal human skin fibroblasts.
Cell biochemistry and function (1991-01-01)
H A Khalfan
ABSTRACT
The protease activity of cultured normal human skin fibroblasts was studied using the synthetic fluorigenic peptides, the modified protein 4-methylumbelliferyl-casein, the thiol inhibitors and the affinity for concanavalin A-Sepharose. The majority of the activity to N-benzyloxycarbonyl-L-phenylalanyl-L-arginyl-7-amido-4-methyl-coumarin and N-a-benzyloxycarbonyl-L-arginyl-arginyl-7-amido-4-methylcoumarin had a pH optimum of 6.0, and was thiol-dependent and inhibited by leupeptin and antipain. The activity toward N-benzyloxycarbonyl-L-phenylalanyl-L-arginyl-7-amido-4-methylcoumarin represents both cathepsin B and cathepsin L, whereas the activity towards 4-methylumbelliferyl-casein represent only cathepsin L. Cathepsin H could not be detected when assayed with L-arginine-7-amido-4-methylcoumarin substrate. Cathepsin D was present in comparatively small amounts when assayed with 4-methylumbelliferyl-casein. Activity towards 4-methylumbelliferyl-casein had pH optima at 3 and 6 and was stimulated by dithiothreitol. A proportion of the activity at pH 6.0 was not dependent on thiols and not inhibited by leupeptin, and had the general characteristics of a carboxyl proteinase. Over 70 per cent of the activity was in the lysosomal fraction and showed structure-linked latency. All the detectable protein emerged from the immobilized concanavalin A column and the fractions eluted by alpha-methyl-D-mannoside were significantly hydrolysed the synthetic peptides. Only that fraction which bound to concanavalin A was active towards 4-methylumbelliferyl-casein. Cathepsin B had no affinity for concanavalin A-Sepharose due to the absence of glycoprotein content, unlike cathepsin L which showed a strong affinity for concanavalin A-Sepharose.
Sigma-Aldrich
Activated Thiol–Sepharose 4B, lyophilized powder