Merck
  • Distinct metabolites for photoreactive L-phenylalanine derivatives in Klebsiella sp. CK6 isolated from rhizosphere of a wild dipterocarp sapling.
Distinct metabolites for photoreactive L-phenylalanine derivatives in Klebsiella sp. CK6 isolated from rhizosphere of a wild dipterocarp sapling.
Molecules (Basel, Switzerland) (2013-07-19)
Lei Wang, Wataru Hisano, Yuta Murai, Munenori Sakurai, Yasuyuki Muto, Haruka Ikemoto, Masashi Okamoto, Takashi Murotani, Reika Isoda, Dongyeop Kim, Yasuko Sakihama, Irnayuli R Sitepu, Yasuyuki Hashidoko, Yasumaru Hatanaka, Makoto Hashimoto
ABSTRACT
Photoaffinity labeling is a reliable analytical method for biological functional analysis. Three major photophores--aryl azide, benzophenone and trifluoromethyldiazirine--are utilized in analysis. Photophore-bearing L-phenylalanine derivatives, which are used for biological functional analysis, were inoculated into a Klebsiella sp. isolated from the rhizosphere of a wild dipterocarp sapling in Central Kalimantan, Indonesia, under nitrogen-limiting conditions. The proportions of metabolites were quite distinct for each photophore. These results indicated that photophores affected substrate recognition in rhizobacterial metabolic pathways, and differential photoaffinity labeling could be achieved using different photophore-containing L-phenylalanine derivatives.
Sigma-Aldrich
H-L-Photo-Phe-OH, ≥98%