Merck
  • MEK mediates v-Src-induced disruption of the actin cytoskeleton via inactivation of the Rho-ROCK-LIM kinase pathway.
MEK mediates v-Src-induced disruption of the actin cytoskeleton via inactivation of the Rho-ROCK-LIM kinase pathway.
The Journal of biological chemistry (2002-05-16)
Geraldine Pawlak, David M Helfman
ABSTRACT
Cellular transformation by v-Src is believed to be caused by aberrant activation of signaling pathways that are normally regulated by cellular Src. Using normal rat kidney cells expressing a temperature-sensitive mutant of v-Src, we examined the role of the Raf/MEK/ERK, phosphatidylinositol 3-kinase/Akt, and Rho pathways in morphological transformation and cytoskeletal changes induced by v-Src. Activation of v-Src elicited a loss of actin stress fibers and focal contacts. A decrease in the phosphorylation level of cofilin was detected upon v-Src activation, which is indicative of attenuated Rho function. Inhibition of MEK using U0126 prevented v-Src-induced disruption of the cytoskeleton as well as dephosphorylation of cofilin, whereas treatment with a phosphatidylinositol 3-kinase inhibitor had no protective effect. In normal rat kidney cells stably transformed by v-Src, we found that the chronic activation of MEK induces down-regulation of ROCK expression, thereby uncoupling Rho from stress fiber formation. Taken together, these results establish MEK as an effector of v-Src-induced cytoskeleton disruption, participating in v-Src-induced antagonism of the cellular function of Rho.
Sigma-Aldrich
Rho Activation Assay Kit, PP1/PP2A Toolbox for the selective in vitro dephosphorylation of proteins.