Merck
  • Purification and characterization of beta-N-acetylhexosaminidases and beta-galactosidase from Streptococcus 6646 K.
Purification and characterization of beta-N-acetylhexosaminidases and beta-galactosidase from Streptococcus 6646 K.
Journal of biochemistry (1976-07-01)
T Kiyohara, T Terao, K Shioiri-Nakano, T Osawa
PMID9384
ABSTRACT
Three beta-N-acetylhexosaminidases [EC 3.2.1.52] and one beta-galactosidase [EC 3.2.1.23] were purified from the culture filtrate of streptococcus 6646 group K by a combination of column chromatographies on p-aminophenyl beta-D-thiogalactopyranoside-substituted Sepharose and N-(paminophenyl)oxamic acid-substituted Sepharose. These beta-N-acetylhexosaminidases showed optimal activities between pH 5.0 and 5.5 and could hydrolyze synthetic and glycopeptidic substrates. Glycolipids such as GM2, asialo-GM2, and globoside I were no susceptible to these beta-hexosaminidases. beta-Galactosidase, which was purified more than 11,000-fold, had a substrate specificity rather similar to that of beta-galactosidase from E. coli. This enzyme was inhibited by EDTA and activated by Mn2+, Ca2+, and Mg2+. Problems pertinent to the application of affinity chromatography to the purification of glycosidases are also discussed.
Sigma-Aldrich
N-(p-Aminophenyl)oxamic acid–Agarose, saline suspension
Pricing and availability is not currently available.