Merck
  • Novel ganglioside antigen identified by B cells in human medullary breast carcinomas: the proof of principle concerning the tumor-infiltrating B lymphocytes.
Novel ganglioside antigen identified by B cells in human medullary breast carcinomas: the proof of principle concerning the tumor-infiltrating B lymphocytes.
Journal of immunology (Baltimore, Md. : 1950) (2005-08-06)
Beatrix Kotlan, Peter Simsa, Jean-Luc Teillaud, Wolf Herman Fridman, Jozsef Toth, Michael McKnight, Mark C Glassy
ABSTRACT
The potential tumor-recognizing capacity of B cells infiltrating human breast carcinoma is an important aspect of breast cancer biology. As an experimental system, we used human medullary breast carcinoma because of its heavy B lymphocytic infiltration paralleled to a relatively better prognosis. Ig-rearranged V region V(H)-J(H), Vkappa-Jkappa, and Vlambda-Jlambda genes, amplified by RT-PCR of the infiltrating B cells, were cloned, sequenced, and subjected to a comparative DNA analysis. A combinatorial single-chain variable fragment Ab minilibrary was constructed out of randomly selected V(H) and Vkappa clones and tested for binding activity. Our data analysis revealed that some of the V(H)-J(H), Vkappa-Jkappa, and Vlambda-Jlambda region sequences were being assigned to clusters with oligoclonal predominance, while other characteristics of the Ab repertoire were defined also. A tumor-restricted binder clone could be selected out of the single-chain variable fragment kappa minilibrary tested against membrane fractions of primary breast tumor cells and tumor cell lines, the V(H) of which proved to be the overexpressed V(H)3-1 cluster. The specific binding was confirmed by FACS analysis with primary breast carcinoma cells and MDA-MB 231 cell line. ELISA and thin layer chromatography dot-blot experiments showed this target Ag to be a ganglioside D3 (GD3). Our results are a proof of principle about the capacity of B cells infiltrating breast carcinomas to reveal key cancer-related Ags, such as the GD3. GD3-specific Abs may influence tumor cell progression and could be used for further development of diagnostic and/or therapeutic purposes.