Merck
  • PLZF/ZBTB16, a glucocorticoid response gene in acute lymphoblastic leukemia, interferes with glucocorticoid-induced apoptosis.
PLZF/ZBTB16, a glucocorticoid response gene in acute lymphoblastic leukemia, interferes with glucocorticoid-induced apoptosis.
The Journal of steroid biochemistry and molecular biology (2010-05-04)
Muhammad Wasim, Michela Carlet, Muhammad Mansha, Richard Greil, Christian Ploner, Alexander Trockenbacher, Johannes Rainer, Reinhard Kofler
ABSTRACT
Glucocorticoids (GCs) cause cell cycle arrest and apoptosis in lymphoid cells which is exploited to treat lymphoid malignancies. The mechanisms of these anti-leukemic GC effects are, however, poorly understood. We previously defined a list of GC-regulated genes by expression profiling in children with acute lymphoblastic leukemia (ALL) during systemic GC monotherapy and in experimental systems of GC-induced apoptosis. PLZF/ZBTB16, a transcriptional repressor, was one of the most promising candidates derived from this screen. To investigate its role in the anti-leukemic GC effects, we performed overexpression and knock-down experiments in CCRF-CEM childhood ALL cells. Transgenic PLZF/ZBTB16 alone had no detectable effect on cell proliferation or survival, but reduced sensitivity to GC-induced apoptosis but not apoptosis induced by antibodies against Fas/CD95 or 3 different chemotherapeutics. Knock-down of ZBTB16 entailed a small, but significant, increase in cell death induction by GC. Affymetrix Exon array-based whole genome expression profiling revealed that PLZF/ZBTB16 induction did not significantly alter the expression profile, however, it interfered with the regulation of numerous GC response genes, including BCL2L11/Bim, which has previously been shown to be responsible for cell death induction in CCRF-CEM cells. Thus, the protective effect of PLZF/ZBTB16 can be attributed to interference with transcriptional regulation by GC.