- Senescence bypass screen identifies TBX2, which represses Cdkn2a (p19(ARF)) and is amplified in a subset of human breast cancers.
Senescence bypass screen identifies TBX2, which represses Cdkn2a (p19(ARF)) and is amplified in a subset of human breast cancers.
Nature genetics (2000-11-04)
J J Jacobs, P Keblusek, E Robanus-Maandag, P Kristel, M Lingbeek, P M Nederlof, T van Welsem, M J van de Vijver, E Y Koh, G Q Daley, M van Lohuizen
PMID11062467
ABSTRACT
To identify new immortalizing genes with potential roles in tumorigenesis, we performed a genetic screen aimed to bypass the rapid and tight senescence arrest of primary fibroblasts deficient for the oncogene Bmi1. We identified the T-box member TBX2 as a potent immortalizing gene that acts by downregulating Cdkn2a (p19(ARF)). TBX2 represses the Cdkn2a (p19(ARF)) promoter and attenuates E2F1, Myc or HRAS-mediated induction of Cdkn2a (p19(ARF)). We found TBX2 to be amplified in a subset of primary human breast cancers, indicating that it might contribute to breast cancer development.