Merck
  • Effects of 3-butenyl isothiocyanate on phenotypically different prostate cancer cells.
Effects of 3-butenyl isothiocyanate on phenotypically different prostate cancer cells.
International journal of oncology (2018-09-19)
M J Núñez-Iglesias, S Novio, C García-Santiago, M E Cartea, P Soengas, P Velasco, M Freire-Garabal
ABSTRACT
Isothiocyanates (ITCs) have gained increasing attention since they have been attributed the merits for the potential beneficial effects of cruciferous vegetable dietary consumption on cancer. The aim of the present study was to determine the cytotoxic effects of 3-butenyl ITC (3-BI) on prostate cancer (PC) cells under in vitro conditions. Two androgen-insensitive human PC cell lines, PC-3 and DU145, were assayed. Cells were cultured in the presence of increasing concentrations of 3-BI (5, 10, 30 and 50 µM) in the absence or presence of the chemotherapeutic drug docetaxel (DOCE) (1 and 2 nM). The cytotoxic effects of these compounds were analyzed using the trypan blue exclusion assay at 24, 48 and 72 h. Apoptosis and migration assays were also performed. The results showed that 3-BI induced a dose-dependent cytotoxic effect on PC-3 cells at 24, 48 and 72 h. These effects were significantly higher than those found with DOCE at 72 h of culture. Moreover, 3-BI also potentiated the effects of DOCE in a dose-dependent manner. Additionally, 3-BI showed inhibition of the migration of PC-3 cells. Nevertheless, 3-BI was not effective in the DU145 PC cell line. These results show a promising role for the 3-BI compound as a co-adjuvant agent in DOCE-based therapy in certain types of PC.
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Nunc® Lab-Tek® Chamber Slide system, 4 wells, Permanox® slide, 1.8 cm2/well, sterile, 96/cs