Merck
  • Pharmacological Induction of RAS-GTP Confers RAF Inhibitor Sensitivity in KRAS Mutant Tumors.
Pharmacological Induction of RAS-GTP Confers RAF Inhibitor Sensitivity in KRAS Mutant Tumors.
Cancer cell (2018-10-10)
Ivana Yen, Frances Shanahan, Mark Merchant, Christine Orr, Thomas Hunsaker, Matthew Durk, Hank La, Xiaolin Zhang, Scott E Martin, Eva Lin, John Chan, Yihong Yu, Dhara Amin, Richard M Neve, Amy Gustafson, Avinashnarayan Venkatanarayan, Scott A Foster, Joachim Rudolph, Christiaan Klijn, Shiva Malek
ABSTRACT
Targeting KRAS mutant tumors through inhibition of individual downstream pathways has had limited clinical success. Here we report that RAF inhibitors exhibit little efficacy in KRAS mutant tumors. In combination drug screens, MEK and PI3K inhibitors synergized with pan-RAF inhibitors through an RAS-GTP-dependent mechanism. Broad cell line profiling with RAF/MEK inhibitor combinations revealed synergistic efficacy in KRAS mutant and wild-type tumors, with KRASG13D mutants exhibiting greater synergy versus KRASG12 mutant tumors. Mechanistic studies demonstrate that MEK inhibition induced RAS-GTP levels, RAF dimerization and RAF kinase activity resulting in MEK phosphorylation in synergistic tumor lines regardless of KRAS status. Taken together, our studies uncover a strategy to rewire KRAS mutant tumors to confer sensitivity to RAF kinase inhibition.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Millipore
Protein A Agarose, Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.
Sigma-Aldrich
Anti-Raf-1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-MEK1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
MEK1 Protein, inactive, 50 g, Unactive, N-terminal GST & C-terminal His6-tagged, recombinant full-length human MEK1, for use in Kinase Assays.
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
MAP Kinase 1/Erk1 Protein, inactive, 50 g, Unactive, N-terminal GST-tagged, recombinant, full-length, human MAP Kinase 1/Erk1, for use in Kinase Assays.
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-B-Raf Antibody, Upstate®, from rabbit
Sigma-Aldrich
MAP Kinase 2/Erk2 Protein, inactive, Human, 50 g, Unactive, N-terminal GST-tagged, recombinant human full length MAP Kinase 2, for use in Kinase Assays.