Merck
  • Immune Checkpoint Inhibition Overcomes ADCP-Induced Immunosuppression by Macrophages.
Immune Checkpoint Inhibition Overcomes ADCP-Induced Immunosuppression by Macrophages.
Cell (2018-10-06)
Shicheng Su, Jinghua Zhao, Yue Xing, Xiaoqian Zhang, Jiang Liu, Qian Ouyang, Jianing Chen, Fengxi Su, Qiang Liu, Erwei Song
ABSTRACT
Antibody-dependent cellular cytotoxicity (ADCC) and antibody-dependent cellular phagocytosis (ADCP) critically contribute to the efficacy of anti-tumor therapeutic antibodies. We report here an unexpected finding that macrophages after ADCP inhibit NK cell-mediated ADCC and T cell-mediated cytotoxicity in breast cancers and lymphomas. Mechanistically, AIM2 is recruited to the phagosomes by FcγR signaling following ADCP and activated by sensing the phagocytosed tumor DNAs through the disrupted phagosomal membrane, which subsequently upregulates PD-L1 and IDO and causes immunosuppression. Combined treatment with anti-HER2 antibody and inhibitors of PD-L1 and IDO enhances anti-tumor immunity and anti-HER2 therapeutic efficacy in mouse models. Furthermore, neoadjuvant trastuzumab therapy significantly upregulates PD-L1 and IDO in the tumor-associated macrophages (TAMs) of HER2+ breast cancer patients, correlating with poor trastuzumab response. Collectively, our findings unveil a deleterious role of ADCP macrophages in cancer immunosuppression and suggest that therapeutic antibody plus immune checkpoint blockade may provide synergistic effects in cancer treatment.
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Millipore
Digitonin, High Purity, CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
EdU Flow Cytometry Kit 555, sufficient for 50 assays