Merck
  • Systematic Identification of MCU Modulators by Orthogonal Interspecies Chemical Screening.
Systematic Identification of MCU Modulators by Orthogonal Interspecies Chemical Screening.
Molecular cell (2017-08-19)
Daniela M Arduino, Jennifer Wettmarshausen, Horia Vais, Paloma Navas-Navarro, Yiming Cheng, Anja Leimpek, Zhongming Ma, Alba Delrio-Lorenzo, Andrea Giordano, Cecilia Garcia-Perez, Guillaume Médard, Bernhard Kuster, Javier García-Sancho, Dejana Mokranjac, J Kevin Foskett, M Teresa Alonso, Fabiana Perocchi
ABSTRACT
The mitochondrial calcium uniporter complex is essential for calcium (Ca2+) uptake into mitochondria of all mammalian tissues, where it regulates bioenergetics, cell death, and Ca2+ signal transduction. Despite its involvement in several human diseases, we currently lack pharmacological agents for targeting uniporter activity. Here we introduce a high-throughput assay that selects for human MCU-specific small-molecule modulators in primary drug screens. Using isolated yeast mitochondria, reconstituted with human MCU, its essential regulator EMRE, and aequorin, and exploiting a D-lactate- and mannitol/sucrose-based bioenergetic shunt that greatly minimizes false-positive hits, we identify mitoxantrone out of more than 600 clinically approved drugs as a direct selective inhibitor of human MCU. We validate mitoxantrone in orthogonal mammalian cell-based assays, demonstrating that our screening approach is an effective and robust tool for MCU-specific drug discovery and, more generally, for the identification of compounds that target mitochondrial functions.
Sigma-Aldrich
CGP-37157, ≥98% (HPLC), powder
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
D-myo-Inositol 1,4,5-tris-phosphate trisodium salt, ≥95% (by 1H NMR and TLC)
Sigma-Aldrich
2,5-Di-tert-butyl-1,4-benzoquinone, 99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Ru360, Ru360, is a cell-permeable oxygen-bridged dinuclear ruthenium amine complex. Binds to mitochondria with high affinity (Kd = 340 pM) and blocks Ca2+ uptake into mitochondria in vitro (IC₅₀ = 184 pM).
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Amiodarone hydrochloride, ≥98%