Merck
  • Oleanolic acid and its derivatives: new inhibitor of protein tyrosine phosphatase 1B with cellular activities.
Oleanolic acid and its derivatives: new inhibitor of protein tyrosine phosphatase 1B with cellular activities.
Bioorganic & medicinal chemistry (2008-08-19)
Yi-Nan Zhang, Wei Zhang, Di Hong, Lei Shi, Qiang Shen, Jing-Ya Li, Jia Li, Li-Hong Hu
ABSTRACT
Protein tyrosine phosphatase 1B is a key factor in the negative regulation of insulin pathway and a promising target for treatment of diabetes and obesity. Herein, a series of competitive inhibitors were optimized from oleanolic acid, a natural triterpenoid identified against PTP1B by screening libraries of traditional Chinese medicinal herbs. Modifying at 3 and 28 positions, we obtained compound 13 with a K(i) of 130 nM, which exhibited good selectivity between other phosphatases involved in insulin pathway except T-cell protein tyrosine phosphatase. Further evaluation in cell models illustrated that the derivatives enhanced insulin receptor phosphorylation in CHO/hIR cells and also stimulated glucose uptake in L6 myotubes with or addition of without insulin.
Sigma-Aldrich
Ursolic acid, ≥90%
Pricing and availability is not currently available.
Sigma-Aldrich
Oleanolic acid, ≥97%
Pricing and availability is not currently available.
Supelco
Ursolic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%
Pricing and availability is not currently available.