Merck
  • Abnormal spermatogenesis and male infertility in testicular zinc finger protein Zfp318-knockout mice.
Abnormal spermatogenesis and male infertility in testicular zinc finger protein Zfp318-knockout mice.
Development, growth & differentiation (2016-07-08)
Masamichi Ishizuka, Eri Ohtsuka, Atsuto Inoue, Mirei Odaka, Hirotaka Ohshima, Norihisa Tamura, Kaoru Yoshida, Norihisa Sako, Tadashi Baba, Shin-Ichi Kashiwabara, Masaru Okabe, Junko Noguchi, Hiromi Hagiwara
ABSTRACT
Zfp318, a mouse gene with a Cys2/His2 zinc finger motif, is mainly expressed in germ cells in the testis. It encodes two alternative transcripts, which regulate androgen receptor-mediated transcriptional activation or repression by overexpression of them. However, the role of Zfp318 is still obscure in vivo, especially in spermatogenesis. To elucidate the role of Zfp318 during gamete production, we established a knockout mouse line. Zfp318-null male mice exhibited infertility, whereas Zfp318-null female mice displayed normal fertility. ZFP318 was expressed during multiple stages of spermatogenesis, from spermatocytes to round spermatids. The nuclei of secondary spermatocytes showed high levels of expression. Histological analysis and quantitative analysis of DNA content showed decreased numbers of both spermatids in the seminiferous tubules and mature spermatozoa in the epididymides of Zfp318-null mice. These results suggest that Zfp318 is expressed as a functional protein in testicular germ cells and plays an important role in meiosis during spermatogenesis.
Sigma-Aldrich
Anti-ZNF318 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Pricing and availability is not currently available.