Merck
  • Fatty acid elongation in yeast--biochemical characteristics of the enzyme system and isolation of elongation-defective mutants.
Fatty acid elongation in yeast--biochemical characteristics of the enzyme system and isolation of elongation-defective mutants.
European journal of biochemistry (1998-04-18)
F Dittrich, D Zajonc, K Hühne, U Hoja, A Ekici, E Greiner, H Klein, J Hofmann, J J Bessoule, P Sperling, E Schweizer
ABSTRACT
Elongation of long-chain fatty acids was investigated in yeast mutants lacking endogenous de novo fatty acid synthesis. In this background, in vitro fatty acid elongation was dependent strictly on the substrates malonyl-CoA, NADPH and a medium-chain or long-chain acyl-CoA primer of 10 or more carbon atoms. Maximal activity was observed with primers containing 12-14 carbon atoms, while shorter-chain-length acyl-CoA were almost (octanoyl-CoA) or completely (hexanoyl-CoA, acetyl-CoA) inactive. In particular, acetyl-CoA was inactive as a primer and as extender unit. The Michaelis constants for octanoyl-CoA (0.33 mM), decanoyl-CoA (0.83 mM) lauroyl-CoA (0.05 mM), myristoyl-CoA (0.4 mM) and palmitoyl-CoA (0.13 mM) were determined and were comparable for fatty acid synthesis and elongation. In contrast, the affinity of malonyl-CoA was 17-fold lower for elongation (Km = 0.13 mM) than for the fatty acid synthase (FAS) system. With increasing chain length of the primer (> or = 12:0), fatty acid elongation becomes increasingly sensitive to substrate inhibition. Due to the activation of endogenous fatty acids, ATP exhibits a stimulatory effect at suboptimal but not at saturating substrate concentrations. In the yeast cell homogenate, the specific activity of fatty acid elongation is about 10-20-fold lower than that of de novo fatty acid synthesis. The same elongation activity is observed in respiratory competent and in mitochondrially defective cells. The products of in vitro fatty acid elongation are fatty acids of 15-17 or 22-26 carbon atoms, depending on whether tridecanoyl-CoA or stearoyl-CoA is used as a primer. In vitro, the elongation products are converted in part, by alpha-oxidation, to their odd-chain-length lower homologues or are hydrolyzed to fatty acids. In contrast, no odd-chain-length elongation products or very-long-chain fatty acids (VLCFA) shorter than 26:0 are observed in vivo. Hence, VLCFA synthesis exhibits a higher processivity in vivo than in the cell homogenate. In addition, the in vivo process appears to be protected against side reactions such as hydrolysis or alpha-oxidation. Yeast mutants defective in 12:0 or 13:0 elongation were derived from fas-mutant strains according to their failure to grow on 13:0-supplemented media. In vivo, 12:0 elongation was reduced to 0-10% of the normal level, while 16:0 elongation and VLCFA synthesis were unimpaired. It is concluded that yeast contains either two different elongation systems, or that the respective mutation interferes differentially with medium-chain and long-chain fatty acid elongation. The yeast gene affected in the elongation-defective mutants was isolated and, upon sequencing, identified as the known ELO1 sequence. It encodes a putative membrane protein of 32-kDa molecular mass with no obvious similarity to any of the known FAS component enzymes.
Sigma-Aldrich
Myristoyl coenzyme A lithium salt, ≥80.0%