Merck
  • Mutations in Disordered Regions Can Cause Disease by Creating Dileucine Motifs.
Mutations in Disordered Regions Can Cause Disease by Creating Dileucine Motifs.
Cell (2018-09-11)
Katrina Meyer, Marieluise Kirchner, Bora Uyar, Jing-Yuan Cheng, Giulia Russo, Luis R Hernandez-Miranda, Anna Szymborska, Henrik Zauber, Ina-Maria Rudolph, Thomas E Willnow, Altuna Akalin, Volker Haucke, Holger Gerhardt, Carmen Birchmeier, Ralf Kühn, Michael Krauss, Sebastian Diecke, Juan M Pascual, Matthias Selbach
ABSTRACT
Many disease-causing missense mutations affect intrinsically disordered regions (IDRs) of proteins, but the molecular mechanism of their pathogenicity is enigmatic. Here, we employ a peptide-based proteomic screen to investigate the impact of mutations in IDRs on protein-protein interactions. We find that mutations in disordered cytosolic regions of three transmembrane proteins (GLUT1, ITPR1, and CACNA1H) lead to an increased clathrin binding. All three mutations create dileucine motifs known to mediate clathrin-dependent trafficking. Follow-up experiments on GLUT1 (SLC2A1), the glucose transporter causative of GLUT1 deficiency syndrome, revealed that the mutated protein mislocalizes to intracellular compartments. Mutant GLUT1 interacts with adaptor proteins (APs) in vitro, and knocking down AP-2 reverts the cellular mislocalization and restores glucose transport. A systematic analysis of other known disease-causing variants revealed a significant and specific overrepresentation of gained dileucine motifs in structurally disordered cytosolic domains of transmembrane proteins. Thus, several mutations in disordered regions appear to cause "dileucineopathies."
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
L-Arginine-13C6 hydrochloride, 99 atom % 13C, 95% (CP)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-GLUT-1 Antibody, CT, from rabbit, purified by affinity chromatography
Sigma-Aldrich
L-Arginine-13C6,15N4 hydrochloride, 99 atom % 13C, 99 atom % 15N, 95% (CP)