Merck
  • cGMP modulates basal and activated microvessel permeability independently of [Ca2+]i.
cGMP modulates basal and activated microvessel permeability independently of [Ca2+]i.
The American journal of physiology (1998-06-25)
P He, M Zeng, F E Curry
ABSTRACT
To investigate the mechanisms whereby guanosine 3',5'-cyclic monophosphate (cGMP) modulates microvessel permeability in vivo, we measured changes in microvessel hydraulic conductivity (Lp) and endothelial cytoplasmic Ca2+ concentration ([Ca2+]i) in response to the cGMP analogs 8-bromo-cGMP (8-BrcGMP) and 8-(p-chlorophenylthio)cGMP (8-pCPT-cGMP) in the presence and absence of inflammatory stimuli in intact individually perfused microvessels in frog and rat mesenteries. The cGMP analog caused a transient increase in Lp and potentiated ATP or bradykinin-induced increases in Lp in frog and rat mesenteric microvessels, respectively. The mean peak value of the test Lp/control Lp after exposure to 8-BrcGMP was 5.3 +/- 0.5 in frog microvessels and 2.8 +/- 0.4 in rat microvessels. The ATP-induced increase in Lp in frog microvessels was further raised by 8-BrcGMP from 7.0 +/- 0.9 to 12.4 +/- 1.9 times the control. In rat mesenteric microvessels, the bradykinin-induced increase in Lp was potentiated by 8-BrcGMP from 4.8 +/- 0.4 to 8.3 +/- 1.3 times the control and was suppressed by the guanylate cyclase inhibitor LY-83583 to 2.6 +/- 0.5 times the control. A similar but larger effect was found when using 8-pCPT-cGMP. In contrast to the actions of increased cGMP on microvessel permeability, cGMP analogs had no effect on basal endothelial [Ca2+]i and did not alter the magnitude and time course of ATP or bradykinin-induced increases in endothelial [Ca2+]i. These results suggested that an elevation of cGMP levels in endothelial cells is a necessary step to increase microvessel permeability in intact microvessels, and this regulatory process occurs downstream from Ca2+ influx, which differs from that reported in large-vessel endothelium in culture and in vascular smooth muscle cells. Experiments carried on microvessels in both frog and rat mesenteries provided a direct comparison of the endothelial cell regulatory mechanisms between species.
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt, ≥95% (HPLC), powder