Merck
  • BAK/BAX macropores facilitate mitochondrial herniation and mtDNA efflux during apoptosis.
BAK/BAX macropores facilitate mitochondrial herniation and mtDNA efflux during apoptosis.
Science (New York, N.Y.) (2018-02-24)
Kate McArthur, Lachlan W Whitehead, John M Heddleston, Lucy Li, Benjamin S Padman, Viola Oorschot, Niall D Geoghegan, Stephane Chappaz, Sophia Davidson, Hui San Chin, Rachael M Lane, Marija Dramicanin, Tahnee L Saunders, Canny Sugiana, Romina Lessene, Laura D Osellame, Teng-Leong Chew, Grant Dewson, Michael Lazarou, Georg Ramm, Guillaume Lessene, Michael T Ryan, Kelly L Rogers, Mark F van Delft, Benjamin T Kile
ABSTRACT
Mitochondrial apoptosis is mediated by BAK and BAX, two proteins that induce mitochondrial outer membrane permeabilization, leading to cytochrome c release and activation of apoptotic caspases. In the absence of active caspases, mitochondrial DNA (mtDNA) triggers the innate immune cGAS/STING pathway, causing dying cells to secrete type I interferon. How cGAS gains access to mtDNA remains unclear. We used live-cell lattice light-sheet microscopy to examine the mitochondrial network in mouse embryonic fibroblasts. We found that after BAK/BAX activation and cytochrome c loss, the mitochondrial network broke down and large BAK/BAX pores appeared in the outer membrane. These BAK/BAX macropores allowed the inner mitochondrial membrane to herniate into the cytosol, carrying with it mitochondrial matrix components, including the mitochondrial genome. Apoptotic caspases did not prevent herniation but dismantled the dying cell to suppress mtDNA-induced innate immune signaling.
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-VDAC1 Antibody, clone N152B/23, clone N152B/23, from mouse
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid