Merck
  • Effects of antigen removal on a porcine osteochondral xenograft for articular cartilage repair.
Effects of antigen removal on a porcine osteochondral xenograft for articular cartilage repair.
Journal of biomedical materials research. Part A (2018-03-27)
Steve Elder, Hudson Chenault, Paul Gloth, Katie Webb, Ruth Recinos, Emily Wright, Dalton Moran, James Butler, Abdolsamad Borazjani, Avery Cooley
ABSTRACT
Given the limited availability of fresh osteochondral allografts and uncertainty regarding performance of decellularized allografts, this study was undertaken as part of an effort to develop an osteochondral xenograft for articular cartilage repair. The purpose was to evaluate a simple antigen removal procedure based mainly on treatment with SDS and nucleases. Histology demonstrated a preservation of collagenous structure and removal of most nuclei. Immunohistochemistry revealed the apparent retention of α-Gal within osteocyte lacunae unless the tissue underwent an additional α-galactosidase processing step. Cytoplasmic protein was completely removed as shown by Western blot. Quantitatively, the antigen removal protocol was found to extract approximately 90% of DNA from cartilage and bone, and it extracted over 80% of glycosaminoglycan from cartilage. Collagen content was not affected. Mechanical testing of cartilage and bone were performed separately, in addition to testing the cartilage-bone interface, and the main effect of antigen removal was an increase in cartilage hydraulic permeability. In vivo immunogenicity was assessed by subcutaneous implantation into DBA/1 J mice, and the response was typical of a foreign body rather than immune reaction. Thus, an osteochondral xenograft produced as described has the potential for further development into a treatment for osteochondral lesions in the human knee. © 2018 Wiley Periodicals, Inc. J Biomed Mater Res Part A: 106A: 2251-2260, 2018.
Sigma-Aldrich
DNA Quantitation Kit, Fluorescence Assay, Quantitation of DNA using bisBenzimide
Pricing and availability is not currently available.