Merck
  • Synthesis and in vitro screening of novel N-benzyl aplysinopsin analogs as potential anticancer agents.
Synthesis and in vitro screening of novel N-benzyl aplysinopsin analogs as potential anticancer agents.
Bioorganic & medicinal chemistry letters (2011-02-08)
Narsimha Reddy Penthala, Thirupathi Reddy Yerramreddy, Peter A Crooks
ABSTRACT
A series of novel substituted (Z)-5-((1-benzyl-1H-indol-3-yl)methylene)imidazolidin-2,4-diones (3a-f) and (Z)-5-((1-benzyl-1H-indol-3-yl)methylene)-2-iminothiazolidin-4-ones (3g-o) have been synthesized utilizing microwave irradiation. These analogs were evaluated for in vitro cytotoxicity against a panel of 60 human tumor cell lines. Compound 3i exhibits potent growth inhibition against melanoma UACC-257 (GI(50)=13.3 nM) and OVCAR-8 ovarian (GI(50)=19.5 nM) cancer cells while possessing significant cytotoxicity (LC(50)=308 nM and LC(50)=851 nM, respectively) against the same cell lines within this series of compounds. A second analog, 3a, had GI(50) values of 307 and 557 nM against SK-MEL-2 melanoma and A498 renal cancer cell lines, and exhibited GI(50) values ranging from 0.30 to 6 μM against 98% of all cancer cell lines in the 60-cell panel. Thus, (Z)-5-((5-chloro-1-(4-fluorobenzyl)-1H-indol-3-yl)methylene)-2-iminothiazolidin-4-one (3i) and (Z)-methyl 1-(4-cyanobenzyl)-3-((2,5-dioxoimidazolidin-4-ylidene)methyl)-1H-indole-6-carboxylate (3a) can be regarded as useful lead compounds for further structural optimization as antitumor agents.
Sigma-Aldrich
Hydantoin, 98%