Merck
  • Highly enantioselective synthesis of 3-amino-2-oxindole derivatives: catalytic asymmetric alpha-amination of 3-substituted 2-oxindoles with a chiral scandium complex.
Highly enantioselective synthesis of 3-amino-2-oxindole derivatives: catalytic asymmetric alpha-amination of 3-substituted 2-oxindoles with a chiral scandium complex.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-04-22)
Zhigang Yang, Zhen Wang, Sha Bai, Ke Shen, Donghui Chen, Xiaohua Liu, Lili Lin, Xiaoming Feng
ABSTRACT
A highly enantioselective alpha-amination of 3-substituted oxindoles with azodicarboxylates catalyzed by a chiral Sc(OTf)(3)/N,N'-dioxide complex (Tf: triflate) has been developed and affords the corresponding 3-amino-2-oxindole derivatives in high yields (up to 98%) with excellent enantioselectivities (up to 99% ee). The procedure is capable of tolerating a relatively wide range of substrates, and excellent results (92-96% ee) can also be obtained, even in the presence of 0.5 mol % of catalyst loading under mild conditions. These results showed the potential value of the catalytic approach. Moreover, with high synthetic versatility, the product could be easily transformed into optically active 3-amino-3-methyloxindole bearing a chiral quaternary stereogenic center.
Sigma-Aldrich
2-Oxindole, 97%