Merck
  • Nitrothiophene carboxamides, a novel narrow spectrum antibacterial series: Mechanism of action and Efficacy.
Nitrothiophene carboxamides, a novel narrow spectrum antibacterial series: Mechanism of action and Efficacy.
Scientific reports (2018-05-10)
Shahul Hameed P, Nagakumar Bharatham, Nainesh Katagihallimath, Sreevalli Sharma, Radha Nandishaiah, Anirudh P Shanbhag, Teby Thomas, Riya Narjari, Maitrayee Sarma, Purnendu Bhowmik, Prakruthi Amar, Rajani Ravishankar, Ramesh Jayaraman, Kubendran Muthan, Ramesh Subbiah, Vasanthi Ramachandran, V Balasubramanian, Santanu Datta
ABSTRACT
The mechanism of efflux is a tour-de-force in the bacterial armoury that has thwarted the development of novel antibiotics. We report the discovery of a novel chemical series with potent antibacterial properties that was engineered to overcome efflux liability. Compounds liable to efflux specifically via the Resistance Nodulation and cell Division (RND) pump, AcrAB-TolC were chosen for a hit to lead progression. Using structure-based design, the compounds were optimised to lose their binding to the efflux pump, thereby making them potent on wild-type bacteria. We discovered these compounds to be pro-drugs that require activation in E. coli by specific bacterial nitroreductases NfsA and NfsB. Hit to lead chemistry led to the generation of compounds that were potent on wild-type and multi-drug resistant clinical isolates of E. coli, Shigella spp., and Salmonella spp. These compounds are bactericidal and efficacious in a mouse thigh infection model.
Sigma-Aldrich
2-Amino-4-(3,4-difluorophenyl)thiazole, 97%