Merck
  • Dendritic Integration of Sensory Evidence in Perceptual Decision-Making.
Dendritic Integration of Sensory Evidence in Perceptual Decision-Making.
Cell (2018-05-01)
Lukas N Groschner, Laura Chan Wah Hak, Rafal Bogacz, Shamik DasGupta, Gero Miesenböck
ABSTRACT
Perceptual decisions require the accumulation of sensory information to a response criterion. Most accounts of how the brain performs this process of temporal integration have focused on evolving patterns of spiking activity. We report that subthreshold changes in membrane voltage can represent accumulating evidence before a choice. αβ core Kenyon cells (αβc KCs) in the mushroom bodies of fruit flies integrate odor-evoked synaptic inputs to action potential threshold at timescales matching the speed of olfactory discrimination. The forkhead box P transcription factor (FoxP) sets neuronal integration and behavioral decision times by controlling the abundance of the voltage-gated potassium channel Shal (KV4) in αβc KC dendrites. αβc KCs thus tailor, through a particular constellation of biophysical properties, the generic process of synaptic integration to the demands of sequential sampling.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Millipore
Universal Food pathogen ID Membrane, light pink colored, sterile, diameter 70 mm, suitable for microbiology
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%