Merck
  • Dopamine D3 receptor blockade rescues hyper-dopamine activity-induced deficit in novel object recognition memory.
Dopamine D3 receptor blockade rescues hyper-dopamine activity-induced deficit in novel object recognition memory.
Neuropharmacology (2018-02-07)
Pi-Kai Chang, Lung Yu, Jin-Chung Chen
ABSTRACT
Patients afflicted with bipolar disorder demonstrate significant impairments in recognition and episodic memory during acute depressive and manic episodes. These impairments and the related pathophysiology may result from over-activation of the brain dopamine (DA) system. In order to model overactive DA transmission in a well-established novel object recognition (NOR) memory test, we used DA transporter knockdown (DAT-KD) mice, which exhibit reduced DAT expression and display hyper-dopaminergic phenotypes. DAT-KD mice exhibited impaired NOR memory compared to wild-type (WT) mice. This impairment was prevented by administration of FAUC365, a DA D3 receptor (D3R) selective antagonist, prior to object learning. Similarly, D3R knockout (KO)/DAT-KD double mutant mice displayed performance in the NOR test that was comparable to WT mice, suggesting that deficiencies in NOR performance in DAT-KD mice can be compensated by diminishing D3R signaling. GBR12909, a DAT blocker, also impaired NOR performance in WT mice, but not in D3R KO mice. Impaired NOR performance in GBR12909-treated WT mice was also prevented by pretreatment with FAUC365. Together, these findings indicate that reduced DAT activity can impair recognition memory in the NOR test, and D3R appears to be necessary to mediate this effect.
Sigma-Aldrich
L-741,626, ≥98% (HPLC)