Merck
  • Long-term resistance exercise-induced muscular hypertrophy is associated with autophagy modulation in rats.
Long-term resistance exercise-induced muscular hypertrophy is associated with autophagy modulation in rats.
The journal of physiological sciences : JPS (2017-02-19)
Insu Kwon, Yongchul Jang, Joon-Yong Cho, Young C Jang, Youngil Lee
ABSTRACT
Elevation of anabolism and concurrent suppression of catabolism are critical metabolic adaptations for muscular hypertrophy in response to resistance exercise (RE). Here, we investigated if RE-induced muscular hypertrophy is acquired by modulating a critical catabolic process autophagy. Male Wistar Hannover rats (14 weeks old) were randomly assigned to either sedentary control (SC, n = 10) or resistance exercise (RE, n = 10). RE elicited significant hypertrophy of flexor digitorum profundus (FDP) muscles in parallel with enhancement in anabolic signaling pathways (phosphorylation of AKT, mTOR, and p70S6K). Importantly, RE-treated FDP muscle exhibited a significant decline in autophagy evidenced by diminished phosphorylation levels of AMPK, a decrease in LC3-II/LC3-I ratio, an increase in p62 level, and a decline in active form of lysosomal protease CATHEPSIN L in the absence of alterations of key autophagy proteins: ULK1 phosphorylation, BECLIN1, and BNIP3. Our study suggests that RE-induced hypertrophy is achieved by potentiating anabolism and restricting autophagy-induced catabolism.
Sigma-Aldrich
Anti-phospho-ULK1 (Ser555) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Eosin Y Solution, Alcoholic, with Phloxine