Merck
  • Assessing the mechanism and therapeutic potential of modulators of the human Mediator complex-associated protein kinases.
Assessing the mechanism and therapeutic potential of modulators of the human Mediator complex-associated protein kinases.
eLife (2016-12-10)
Paul A Clarke, Maria-Jesus Ortiz-Ruiz, Robert TePoele, Olajumoke Adeniji-Popoola, Gary Box, Will Court, Stephanie Czasch, Samer El Bawab, Christina Esdar, Ken Ewan, Sharon Gowan, Alexis De Haven Brandon, Phillip Hewitt, Stephen M Hobbs, Wolfgang Kaufmann, Aurélie Mallinger, Florence Raynaud, Toby Roe, Felix Rohdich, Kai Schiemann, Stephanie Simon, Richard Schneider, Melanie Valenti, Stefan Weigt, Julian Blagg, Andree Blaukat, Trevor C Dale, Suzanne A Eccles, Stefan Hecht, Klaus Urbahns, Paul Workman, Dirk Wienke
ABSTRACT
Mediator-associated kinases CDK8/19 are context-dependent drivers or suppressors of tumorigenesis. Their inhibition is predicted to have pleiotropic effects, but it is unclear whether this will impact on the clinical utility of CDK8/19 inhibitors. We discovered two series of potent chemical probes with high selectivity for CDK8/19. Despite pharmacodynamic evidence for robust on-target activity, the compounds exhibited modest, though significant, efficacy against human tumor lines and patient-derived xenografts. Altered gene expression was consistent with CDK8/19 inhibition, including profiles associated with super-enhancers, immune and inflammatory responses and stem cell function. In a mouse model expressing oncogenic beta-catenin, treatment shifted cells within hyperplastic intestinal crypts from a stem cell to a transit amplifying phenotype. In two species, neither probe was tolerated at therapeutically-relevant exposures. The complex nature of the toxicity observed with two structurally-differentiated chemical series is consistent with on-target effects posing significant challenges to the clinical development of CDK8/19 inhibitors.