Merck
  • Targeting G protein-coupled receptor signaling at the G protein level with a selective nanobody inhibitor.
Targeting G protein-coupled receptor signaling at the G protein level with a selective nanobody inhibitor.
Nature communications (2018-05-20)
Sahil Gulati, Hui Jin, Ikuo Masuho, Tivadar Orban, Yuan Cai, Els Pardon, Kirill A Martemyanov, Philip D Kiser, Phoebe L Stewart, Christopher P Ford, Jan Steyaert, Krzysztof Palczewski
ABSTRACT
G protein-coupled receptors (GPCRs) activate heterotrimeric G proteins by mediating a GDP to GTP exchange in the Gα subunit. This leads to dissociation of the heterotrimer into Gα-GTP and Gβγ dimer. The Gα-GTP and Gβγ dimer each regulate a variety of downstream pathways to control various aspects of human physiology. Dysregulated Gβγ-signaling is a central element of various neurological and cancer-related anomalies. However, Gβγ also serves as a negative regulator of Gα that is essential for G protein inactivation, and thus has the potential for numerous side effects when targeted therapeutically. Here we report a llama-derived nanobody (Nb5) that binds tightly to the Gβγ dimer. Nb5 responds to all combinations of β-subtypes and γ-subtypes and competes with other Gβγ-regulatory proteins for a common binding site on the Gβγ dimer. Despite its inhibitory effect on Gβγ-mediated signaling, Nb5 has no effect on Gαq-mediated and Gαs-mediated signaling events in living cells.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Apelin Receptor/APJ, clone 3C3-7 Antibody, clone 3C3-7, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse