Merck
  • PD-L1 inhibits acute and chronic pain by suppressing nociceptive neuron activity via PD-1.
PD-L1 inhibits acute and chronic pain by suppressing nociceptive neuron activity via PD-1.
Nature neuroscience (2017-05-23)
Gang Chen, Yong Ho Kim, Hui Li, Hao Luo, Da-Lu Liu, Zhi-Jun Zhang, Mark Lay, Wonseok Chang, Yu-Qiu Zhang, Ru-Rong Ji
ABSTRACT
Programmed cell death ligand-1 (PD-L1) is typically produced by cancer cells and suppresses immunity through the receptor PD-1 expressed on T cells. However, the role of PD-L1 and PD-1 in regulating pain and neuronal function is unclear. Here we report that both melanoma and normal neural tissues including dorsal root ganglion (DRG) produce PD-L1 that can potently inhibit acute and chronic pain. Intraplantar injection of PD-L1 evoked analgesia in naive mice via PD-1, whereas PD-L1 neutralization or PD-1 blockade induced mechanical allodynia. Mice lacking Pd1 (Pdcd1) exhibited thermal and mechanical hypersensitivity. PD-1 activation in DRG nociceptive neurons by PD-L1 induced phosphorylation of the tyrosine phosphatase SHP-1, inhibited sodium channels and caused hyperpolarization through activation of TREK2 K+ channels. PD-L1 also potently suppressed nociceptive neuron excitability in human DRGs. Notably, blocking PD-L1 or PD-1 elicited spontaneous pain and allodynia in melanoma-bearing mice. Our findings identify a previously unrecognized role of PD-L1 as an endogenous pain inhibitor and a neuromodulator.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-PD-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium Stibogluconate, A pentavalent antimony compound that irreversibly inhibits protein tyrosine phosphatase (PTPase) activity, including Src homology PTPase-1 (SHP-1) (99% inhibition at ~11 µM) by forming a stable complex.