Merck
  • Glioblastoma-derived cells in vitro unveil the spectrum of drug resistance capability - comparative study of tumour chemosensitivity in different culture systems.
Glioblastoma-derived cells in vitro unveil the spectrum of drug resistance capability - comparative study of tumour chemosensitivity in different culture systems.
Bioscience reports (2017-05-20)
Monika Witusik-Perkowska, Magdalena Zakrzewska, Beata Sikorska, Wielislaw Papierz, Dariusz J Jaskolski, Janusz Szemraj, Pawel P Liberski
ABSTRACT
Resistance to cancer drugs is a complex phenomenon which could be influenced by in vitro conditions. However, tumour-derived cell cultures are routinely used for studies related to mechanisms of drug responsiveness or the search for new therapeutic approaches. The purpose of our work was to identify the potential differences in drug resistance and response to treatment of glioblastoma with the use of three in vitro models: traditional adherent culture, serum-free spheroid culture and novel adherent serum-free culture.The experimental models were evaluated according to 'stemness state' and epithelial-to-mesenchymal transition (EMT) status, invasion capability and their expression pattern of genes related to the phenomenon of tumour drug resistance. Additionally, the response to drug treatments of three different culture models was compared with regard to the type of cell death.Multi-gene expression profiling revealed differences between examined culture types with regard to the expression pattern of the selected genes. Functionally, the examined genes were related to drug resistance and metabolism, DNA damage and repair and cell cycle control, and included potential therapeutic targets.Cytotoxicity analyses confirmed that environmental factors can influence not only the molecular background of glioblastoma drug-resistance and efficiency of treatment, but also the mechanisms/pathways of cell death, which was reflected by a distinct intensification of apoptosis and autophagy observed in particular culture models. Our results suggest that parallel exploitation of different in vitro experimental models can be used to reveal the spectrum of cancer cell resistance capability, especially regarding intra-heterogeneous glioblastomas.
Sigma-Aldrich
Monoclonal Anti-IL13RA2 antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Twist1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Vimentin antibody, Mouse monoclonal, clone V9, purified from hybridoma cell culture
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-E-cadherin antibody produced in rabbit, affinity isolated antibody