Merck
  • Coordinate suppression of Sdpr and Fhl1 expression in tumors of the breast, kidney, and prostate.
Coordinate suppression of Sdpr and Fhl1 expression in tumors of the breast, kidney, and prostate.
Cancer science (2008-04-22)
Xun Li, Zhenyu Jia, Yongquan Shen, Hitoshi Ichikawa, Jonathan Jarvik, Robert G Nagele, Gary S Goldberg
ABSTRACT
The Src tyrosine kinase associates with the focal adhesion adaptor protein Cas (Crk-associated substrate) to suppress the expression of potential tumor suppressor genes. For example, Src utilizes Cas to suppress the expression of the LIM-only protein Fhl1 (four and a half LIM domains 1), in order to promote non-anchored tumor-cell growth and migration. Here, we report that the promoter region of the Fhl1 gene was methylated more in Src-transformed cells than non-transformed cells. In addition, global expression analysis indicates that Fhl1 induced expression of serum deprivation response factor (Sdpr) in Src-transformed cells. Moreover, Fhl1 and Sdpr was expressed in approximately 87% and 40% of samples obtained from non-transformed breast, 100% of samples obtained from non-transformed kidney, and over 60% of samples obtained from non-transformed prostate. In contrast, Fhl1 and Sdpr was detected in approximately 40% and 7% of matched samples from mammary carcinoma, less than 11% of matched samples from kidney carcinoma, and in less than 22% of matched samples from prostate carcinoma. These data indicate that Fhl1 and Sdpr expression was significantly reduced in tumors of the breast (P < 0.02 and P < 0.001), kidney (P < 0.01), and prostate (P < 0.05). In addition, although Src can activate mitogen-activated protein kinase (MAPK) to promote tumor-cell growth, our data indicate that Src did not rely on MAPK activity to suppress the expression of Fhl1 and Sdpr in transformed cells. Thus, Src induced methylation of the promoter region of the Fhl1 gene; Src suppressed Fhl1 and Sdpr expression independent of mitogen-activated protein kinase (MAPK) activity; Fhl1 induced the expression of Sdpr in Src-transformed cells; and Fhl1 and Sdpr expression was suppressed in tumors of the breast, kidney, and prostate.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture