Merck
  • Minodronic Acid in Combination with γδT Cells Induces Apoptosis of Non-small Cell Lung Carcinoma Cell Lines.
Minodronic Acid in Combination with γδT Cells Induces Apoptosis of Non-small Cell Lung Carcinoma Cell Lines.
Anticancer research (2016-10-30)
Motoaki Yasukawa, Tsutomu Nakazawa, Takeshi Kawaguchi, Norikazu Kawai, Takahiro Tsujimura, Takashi Tojo, Shigeki Taniguchi
ABSTRACT
Non-small cell lung carcinoma (NSCLC) is one of the leading causes of cancer-related death worldwide. Recent studies showed that nitrogen-containing bisphosphonates (N-BPs) directly and indirectly prevent proliferation, induce apoptosis, and inhibit metastasis of various types of cancer cell. In order to investigate the effect of combining minodronic acid (MDA) with γδ T-cells, NSCLC cells were treated with five concentrations of MDA. NSCLC cells were cultured with different concentrations of MDA alone or in combination with γδ T-cells for 24 h. MDA with γδ T-cells had differential apoptotic effects on the NSCLC cell lines. The cells showed significant apoptotic effects in the presence of MDA in a dose-dependent manner. This study is the first to report an indirect effect of MDA against NSCLC. We report the induction of apoptosis by MDA in combination with γδ T-cells.
Sigma-Aldrich
Minodronic acid monohydrate, ≥98% (HPLC)