![Z-L-Phe chloromethyl ketone 98%](/deepweb/assets/sigmaaldrich/product/structures/222/435/d4f5b858-132f-4752-9c28-6fcbceb453f7/640/d4f5b858-132f-4752-9c28-6fcbceb453f7.png)
Z-L-Phe chloromethyl ketone
![Z-L-Phe chloromethyl ketone 98%](/deepweb/assets/sigmaaldrich/product/structures/222/435/d4f5b858-132f-4752-9c28-6fcbceb453f7/640/d4f5b858-132f-4752-9c28-6fcbceb453f7.png)
Synonym(s):
N-Carbobenzyloxy-L-phenylalanyl chloromethyl ketone, NSC 251810, SL-01, ZPCK
Linear Formula:
C6H5CH2CH(NHCO2CH2C6H5)COCH2Cl
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing